Learn More
Macherey-Nagel™ Ozone Test Strips
Product Code. 10648192 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Brown
Unit Size:
Pack of 12
Product Code. 10648192
Brand: Macherey-Nagel™ 90736
Specifications
Color Indicator White to Brown
Format Strip
For Use With (Application) Ozone testing
Graduations <90, 90 to 150, 150 to 210, >210μg/m3 O3
Parameter Specifications Ozone
Quantity 12 Pack
Test Method Ozone
Type Test Strip