missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%

Product Code. 10356910 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 10356910

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-64-6
Molecular Formula C22H28N2O11
Molecular Weight (g/mol) 496.469
MDL Number MFCD00151234
InChI Key IMLJLCJZQLGHJS-JEKSYDDFSA-N
PubChem CID 54680691
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
SMILES CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O

Specifications

Type Oxytetracycline Dihydrate
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation On anhydrous substance: -203° to -216° (20°C, 589nm) (c=1, 0.1M HCl)
Water 4 to 8%
Color Yellow
Quantity 100 g
Assay Percent Range 95%
Merck Index 14, 6976
Formula Weight 496.46
Percent Purity ≥94% (on anhydrous substance)
Chemical Name or Material Oxytetracycline dihydrate, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxytetracycline dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.