Learn More
Hirschmann™ Oxygen Bottles
Product Code. 11712293
Change view
Click to view available options
Volume (Metric):
200 to 300 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11712293 200 to 300 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11712293 Supplier Hirschmann™ Supplier No. 6500187
  • ONLINE EXCLUSIVE PRICE
    963.90 NOK
    890.28 NOK / Pack of 2
    Save 73.62 NOK
    7% Off
Estimated Shipment: 10-02-2027
Individually marked and calibrated oxygen bottles acc. To winkler
  • Acc. to Winkler
  • White graduation
  • Precisely adjusted
  • With labelling square
  • Consecutively serial-numbered on the bottle and the bevelled glass stopper
Specifications
Volume (Metric) 200 to 300 mL
Height (Metric) 135 mm