Learn More
Oxyfluorfen, TRC
Product Code. 30135873
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135873 10 g 10g
17975105 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135873 Supplier Toronto Research Chemicals Supplier No. TRCO87075010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42874-03-3
Molecular Formula C15 H11 Cl F3 N O4
Molecular Weight (g/mol) 361.7
Synonym Benzene, 2-chloro-1-(3-ethoxy-4-nitrophenoxy)-4-(trifluoromethyl)-, 2-Chloro-1-(3-ethoxy-4-nitrophenoxy)-4-(trifluoromethyl)benzene, 2-Chloro-4-trifluoromethylphenyl 3-ethoxy-4-nitrophenyl ether, 2'-Chloro-3-ethoxy-4'-trifluoromethyl-4-nitrodiphenyl ether, 4-Trifluoromethyl-2-chloro-3'-ethoxy-4'-nitrophenyl ether, Fluoxil 24EC, Galigan, Goal, Goal 2E, Goal 2XL, Koltar, Oxyfluorfen, Oxygold, RH 2915, RH 2915D
IUPAC Name 2-chloro-1-(3-ethoxy-4-nitrophenoxy)-4-(trifluoromethyl)benzene
SMILES CCOc1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C15H11ClF3NO4/c1-2-23-14-8-10(4-5-12(14)20(21)22)24-13-6-3-9(7-11(13)16)15(17,18)19/h3-8H,2H2,1H3
Formula Weight 361.0329
Percent Purity >95%
Chemical Name or Material Oxyfluorfen
Grade HPLC