Learn More
Oxyfluorfen 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30310682
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310682 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310682 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010369ME
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxyfluorfen
CAS 42874-03-3
Analyte or Component Names Oxyfluorfen
Shipping Condition Room Temperature
Molecular Formula C15 H11 Cl F3 N O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C15H11ClF3NO4/c1-2-23-14-8-10(4-5-12(14)20(21)22)24-13-6-3-9(7-11(13)16)15(17,18)19/h3-8H,2H2,1H3
SMILES CCOc1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]
IUPAC Name 2-chloro-1-(3-ethoxy-4-nitrophenoxy)-4-(trifluoromethyl)benzene
Solvent or Matrix Methanol
Molecular Weight (g/mol) 361.7
Quantity 1 mL
Formula Weight 361.0329 g/mol