Learn More
Thermo Scientific™ Oxoid™ Violet Red Bile Agar (VRB Agar)
Product Code. 15333118
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15333118 10 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15333118 Supplier Thermo Scientific™ Supplier No. PO5075A
  • ONLINE EXCLUSIVE PRICE
    106.00 NOK
    104.18 NOK / Pack of 20
    Save 1.82 NOK
    1% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Enhance the detection of enteric bacteria with this agar, providing selective isolation for food and water testing.
Enhance the detection of enteric bacteria with Violet Red Bile Agar (VRB Agar), providing selective isolation for food and water testing.
TRUSTED_SUSTAINABILITY
Specifications
Color Violet
Content And Storage 2°C to 12°C
Format 90 mm Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17 g
Quantity 10 Plates