missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Vegetable Peptone No. 1

Product Code. 12755650
Click to view available options
Quantity:
500 g
25 kg
Unit Size:
500g
Each
This item is not returnable. View return policy

Product Code. 12755650

Brand: Thermo Scientific™ VG0100K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Oxoid Vegetable Peptone No. 1 is a meat-free general purpose peptone.

Thermo Scientific™ Oxoid Vegetable Peptone No. 1 is a highly nutritious, meat-free general purpose peptone for growth of bacteria and fungi in culture media or fermentation processes.

25 Kg Vegetable Peptone No.1 Oxoid Dehydrated
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 25 kg
Protein Source Pea

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.