Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Product Code. p-4522323
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 12638797
Brand: Thermo Scientific™ SR0247E
Edge
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Oxoid™ Vancomycin Supplement
Form Powder
Product Type Microbiology Media
Quantity 10/Pk.
Species Vancomycin-resistant Enterococci (VRE), High-level Aminoglucoside-resistant Enterococci (HLARE)