Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Oxoid™ Vancomycin Supplement |
| Form | Powder |
| Product Type | Microbiology Media |
| Packaging Type | 5 mg/Vial |
| Quantity | 10/Pk. |
| Species | Vancomycin-resistant Enterococci (VRE), High-level Aminoglucoside-resistant Enterococci (HLARE) |