Learn More
Thermo Scientific™ Oxoid™ Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility discs, 25μg
Click to view available options
Concentration:
25 μg
Unit Size:
Pack of 250
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Antimicrobial Agents | Trimethoprim/Sulfamethoxazole |
| Certifications/Compliance | Complies with CLSI, DIN, SFM guidelines |
| Concentration | 25 μg |
| Description | Oxoid Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility Discs |
| Antibiotic Code | SXT |
| Product Type | Antimicrobial Susceptibility Discs |
| Test Type | Antimicrobial Susceptibility |
| No. per Pack | 5 Cartridges/Pk. |
| Packaging Type | 50 Discs/Cartridge |
| Quantity | 50 Discs/Cartridge |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Documents
Scientific Resources
Brochures
Oxoid Antimicrobial Susceptibility Disks Brochure - Global [EN]
File size: 773 kB
Antimicrobial Susceptibility Testing (AST) Range Brochure
File size: 2 MB
Product Information
Instructions
X7215F CTs Generic IFU - IVDR_ Catenated
File size: 1.1 MB
MBD_BT_IFU-0755 CT0052B AST Discs - TrimethoprimSulfamethoxazole.pdf
File size: 2.3 MB
Specification Sheets
PRODUCT SPECIFICATION:CT0052B SULPHAMETHOXAZOLE/TRIMETHOPRIM SXT25 (V3)
File size: 193.6 kB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
| Lot Number | Certificate Type | Date | Product Code |
|---|---|---|---|
| 6335676 | Certificate of Analysis | 13/11/2025 | CT0052B |
| 6326517 | Certificate of Analysis | 30/10/2025 | CT0052B |
| 6326859 | Certificate of Analysis | 30/10/2025 | CT0052B |
| 6323162 | Certificate of Analysis | 27/10/2025 | CT0052B |
| 6320727 | Certificate of Analysis | 09/10/2025 | CT0052B |
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Trimethoprim/Sulfamethoxazole 1:19 Antimicrobial Susceptibility discs, 25μg >