Learn More
Thermo Scientific™ Oxoid™ Skim Milk Powder
Product Code. 16694685
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16694685 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16694685 Supplier Thermo Scientific™ Supplier No. LP0033B
  • ONLINE EXCLUSIVE PRICE
    535.00 NOK
    464.22 NOK / 500g
    Save 70.78 NOK
    13% Off
 Restricted Item
In Stock
Oxoid Skim Milk Powder is a thermophile-free, spray dried powder for inclusion in bacteriological culture media.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: NMKL.
Description Skim Milk Powder
Form Powder
Product Type Milk
Quantity 500 g