Learn More
Thermo Scientific™ Oxoid™ Proteose Peptone
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Thermo Scientific™ Oxoid Proteose Peptone is a specialty peptone containing proteoses as defined in the USP.
Particularly suitable for use in toxin production and fermentation applications or in culture media for bacteria with wide-ranging nutritional requirements.
- Particularly suitable for use in toxin production and in culture media for bacteria with wide-ranging nutritional requirements
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: USP. |
| Content And Storage | Dehydrated Powder, Ambient & Dry Storage |
| Form | Dehydrated Powder |
| Product Type | Peptone |
| For Use With (Application) | Culture media or fermentation processes |
| Quantity | 500 g |
| Protein Source | Pork / Beef / Soy |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.