Learn More
Thermo Scientific™ Oxoid™ Peptone Mycological
Product Code. p-200112598
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
10630195 500 g 500g
1 options
Product Code. 10630195
Brand: Thermo Scientific™ LP0040B
Oxoid Peptone Mycological is a mycological peptone developed for isolation of pathogenic and nonpathogenic fungi.
TRUSTED_SUSTAINABILITY
Specifications
Description Peptone Mycological
Form Powder
Product Type Peptone
Quantity 500 g