Learn More
Thermo Scientific™ Microbact™ GNB Kit
missing translation for 'orderingAttributeHoverText'
Description:
Microbact GNB 12A Kit
Microbact GNB 12B Kit
Microbact GNB 24E Kit
Quantity:
40 Tests/Kit
60 Tests/Kit
missing translation for 'unitSize'
40 tests
60 tests
Description
Simplify the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram-negative bacteria) with the Thermo Scientific™ Oxoid™ Microbact™ GNB Kit – a complete, self-contained biochemical based identification system. The kit uses 24 different biochemical tests in microplate format to produce easy-to read, distinct color reactions, generally following overnight incubation.
Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With this GNB Kit, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast and appropriate action may follow.
Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The six reagents above are also available as Reagent Set D (MB1082A).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
- Fast: Most results available overnight
- Simple and Economical: Simple microplate format
- Easy-to-Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
- Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
- Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
- Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4
Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Software Program (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The six reagents above are also available as Reagent Set D (MB1082A).
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Spécification
Spécification
| Description | Microbact GNB 12B Kit |
| Detectable Analytes | Gram-negative bacilli |
| Packaging Type | 1 Box |
| Quantity | 60 Tests/Kit |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu