missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Hemoglobin Soluble Powder

Product Code. p-200112603
Click to view available options
Quantity:
1 Each
Unit Size:
400g
This item is not returnable. View return policy

Product Code. 15660421

Brand: Thermo Scientific™ LP0053C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Hemoglobin Powder Soluble is a specially prepared hemoglobin for use in culture media.

Oxoid Hemoglobin Powder Soluble is a specially prepared hemoglobin for use in culture media.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Shipped at room temperature. Store at 10° to 30°C. Keep tightly closed.
Description Dehydrated; Hemoglobin powder
Form Powder
Product Type Hemoglobin
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.