Learn More
Thermo Scientific™ Oxoid™ GMO-Free Soya Peptone
Product Code. p-4524669
Click to view available options
Quantity:
500 g
Unit Size:
400g
Product Code. 15332680
Brand: Thermo Scientific™ VG0300C
Edge
Oxoid GMO-Free Soya Peptone is a nutritious, general-purpose peptone for the growth of bacteria and fungi.
Oxoid GMO-Free Soya Peptone is a nutritious, general-purpose peptone for the growth of bacteria and fungi.
A nutritious, general purpose peptone for the growth of bacteria and fungi.
TRUSTED_SUSTAINABILITY
Specifications
Description GMO-Free Soya Peptone
Form Powder
Product Type Peptone
Quantity 500 g
For Laboratory Use Only