Learn More
Oxiracetam, LoGiCal
Product Code. 30464087
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30464087
Brand: LoGiCal LGCFOR1406.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxiracetam
CAS 62613-82-5
Analyte or Component Names Oxiracetam
Shipping Condition Room Temperature
Molecular Formula C6 H10 N2 O3
Certifications/Compliance ISO 9001
Synonym 2-(4-Hydroxy-2-oxopyrrolidin-1-yl)acetamide, 4-Hydroxy-2-oxo-1-pyrrolidineacetamide, 4-Hydroxypyrrolidone-2-acetamide, CGP 21690, CGP 21690E, CT 848, Hydroxypiracetam, ISF 2522, Neuractiv, Neuromet, Oxiracetam
InChI Formula InChI=1S/C6H10N2O3/c7-5(10)3-8-2-4(9)1-6(8)11/h4,9H,1-3H2,(H2,7,10)
Solution Type Neat
SMILES NC(=O)CN1CC(O)CC1=O
IUPAC Name 2-(4-hydroxy-2-oxopyrrolidin-1-yl)acetamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 158.16
Quantity 10 mg
Formula Weight 158.0691 g/mol