missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OXGR1 Recombinant Rabbit Monoclonal Antibody (23GB4575), Invitrogen™
GREENER_CHOICE

Product Code. p-200088038
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087674 100 μL 100µL
30089018 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087674 Supplier Invitrogen™ Supplier No. MA553772

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXGR1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4575
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OXGR1
Gene Accession No. Q6IYF8, Q6Y1R5, Q96P68
Gene Alias 2-oxoglutarate receptor 1; aKGR; Alpha-ketoglutarate receptor 1; Cysltr3; G protein-coupled receptor 80; G protein-coupled receptor 99; Gm299; Gpr80; Gpr99; G-protein coupled receptor 80; G-protein coupled receptor 99; MGC119206; MGC119207; MGC119208; Oxgr1; oxoglutarate (alpha-ketoglutarate) receptor 1; oxoglutarate receptor 1; P2RY15; P2Y purinoceptor 15; P2Y15; P2Y-like GPCR; P2Y-like nucleotide receptor; seven transmembrane helix receptor
Gene Symbols OXGR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OXGR1/GPR99 (181-215AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239283, 27199, 290493
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.