Learn More
Oxantel Pamoate, TRC
Product Code. 17994235
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994235 1 g 1g
17912185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17994235 Supplier Toronto Research Chemicals Supplier No. TRCO8452501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68813-55-8
Molecular Formula C23 H16 O6 . C13 H16 N2 O
Molecular Weight (g/mol) 604.65
Synonym 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with (E)-3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), Phenol, 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt) (9CI), Phenol, 3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, (E)-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt), CP 14445-16, Oxantel ebonate, Oxantel pamoate, Telopar
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;3-[(E)-2-(1-methyl-5,6-dihydro-4H-pyrimidin-2-yl)ethenyl]phenol
SMILES CN1CCCN=C1\C=C\c2cccc(O)c2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C23H16O6.C13H16N2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-15-9-3-8-14-13(15)7-6-11-4-2-5-12(16)10-11/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-7,10,16H,3,8-9H2,1H3/b;7-6+
Formula Weight 604.221
Chemical Name or Material Oxantel Pamoate