missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, Supelco™

Product Code. 15886850
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Unit Size:
100g
1kg
25kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886850 25 kg 25kg
15833900 100 g 100g
15843900 500 g 500g
15853900 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15886850 Supplier Supelco Supplier No. 1004959025
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6

Specifications

Melting Point 98°C to 100°C
Boiling Point 149°C to 160°C/1013 hPa (decomposition)
Density 1.65 g/cm3 at 20°C
Quantity 25 kg
Molecular Formula (COOH)2 · 2 H2O
MDL Number MFCD00149102
Synonym Oxalic acid dihydrate, Ethanedioic acid, Oxalic acid dihydrate
Molecular Weight (g/mol) 126.07
Grade Ungraded
Chemical Name or Material Oxalic Acid Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.