Learn More
Oxalic Acid Dihydrate, Sigma-Aldrich™
Product Code. 15890311
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15890311 100 g 100g
15806570 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15890311 Supplier Sigma-Aldrich Supplier No. 8182420100
 Restricted Item
Estimated Shipment: 12-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6153-56-6
Specifications
Melting Point 98°C to 100°C
Boiling Point 149°C to 160°C/1013 hPa (decomposition)
Density 1.65 g/cm3 at 20°C
Quantity 100 g
Molecular Formula (COOH)2 · 2 H2O
MDL Number MFCD00149102
Synonym Oxalic acid dihydrate, Ethanedioic acid, Oxalic acid dihydrate
Molecular Weight (g/mol) 126.07
Grade Ungraded
Chemical Name or Material Oxalic Acid Dihydrate