Learn More
Oxacillin sodium monohydrate, MedChemExpress
Product Code. 16458326
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16458326
Brand: Medchem Express HYB0465
MedChemExpress Oxacillin sodium monohydrate is an antibiotic similar to Flucloxacillin used in resistant staphylococci infections study.
Oxacillin sodium monohydrate is an antibiotic similar to Flucloxacillin used in resistant staphylococci infections study.
Specifications
Chemical Name or Material Oxacillin sodium monohydrate
Physical Form Solid
CAS 7240-38-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H20N3NaO6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 100 mg/mL (226.54 mM) ∣DMSO : 50 mg/mL (113.27 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@@H]1N(C2=O)[C@]([C@@H]2NC(C(C(C3=CC=CC=C3)=NO4)=C4C)=O)([H])SC1(C)C.O
Molecular Weight (g/mol) 441.43
Formula Weight 441.43
Percent Purity 98.1%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.