missing translation for 'onlineSavingsMsg'
Learn More

Oxacillin sodium monohydrate, MedChemExpress

Product Code. 16452146
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16452146

Brand: Medchem Express HYB0465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxacillin sodium monohydrate is an antibiotic similar to Flucloxacillin used in resistant staphylococci infections study.

Oxacillin sodium monohydrate is an antibiotic similar to Flucloxacillin used in resistant staphylococci infections study.

Specifications

Chemical Name or Material Oxacillin sodium monohydrate
Physical Form Solid
CAS 7240-38-2
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H20N3NaO6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 100 mg/mL (226.54 mM) ∣DMSO : 50 mg/mL (113.27 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@@H]1N(C2=O)[C@]([C@@H]2NC(C(C(C3=CC=CC=C3)=NO4)=C4C)=O)([H])SC1(C)C.O
Molecular Weight (g/mol) 441.43
Formula Weight 441.43
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.