Learn More
Oxaceprol, MedChemExpress
Product Code. 16401689
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16401689 Supplier Medchem Express Supplier No. HY17490
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Oxaceprol (N-Acetyl-L-hydroxyproline), an orally active derivative of L-proline, possesses distinct anti-inflammatory activity. Oxaceprol is usually used for the research of osteoarthritis and rheumatoid arthritis.
Oxaceprol (N-Acetyl-L-hydroxyproline), an orally active derivative of L-proline, possesses distinct anti-inflammatory activity. Oxaceprol is usually used for the research of osteoarthritis and rheumatoid arthritis.
Specifications
Chemical Name or Material Oxaceprol
CAS 33996-33-7
Color White
Purity Grade Notes Research
Molecular Formula C7H11NO4
Quantity 1 g
Synonym N-Acetyl-L-hydroxyproline
Solubility Information DMSO : ≥ 100 mg/mL (577.47 mM)
SMILES O=C(O)[C@H]1N(C(C)=O)C[C@H](O)C1
Molecular Weight (g/mol) 173.17
Formula Weight 173.17
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation