Learn More
Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts
Product Code. 11520495
Change view
Click to view available options
Description:
Buffer drain bottle
Replacement Knob (Pack of 6)
Side Clamps; 1 pair
Unit Size:
Each
Pack of 6
Product Code. Description unitSize
11520495 Buffer drain bottle Each
12484727 Side Clamps; 1 pair Each
11962985 Replacement Knob (Pack of 6) Pack of 6
3 options
Product Code. 11520495
Brand: Thermo Scientific™ R10383
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts.
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S3S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffer drain bottle
For Use With (Equipment) Owl™ S3S Aluminum-Backed Sequencer System
Product Type Drain Bottle