Learn More
Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster
Product Code. 12494687
Click to view available options
Type:
Caster
Frosted Blank Glass
Frosted Notched Glass
Unit Size:
Each
Product Code. 12494687
Brand: Thermo Scientific™ P710FG
Only null left
Add to basket
Add to basket
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.
Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P7-CST System
Product Line Owl
Type Frosted Blank Glass
Thickness (English) 0.09 in.
Length (Metric) 10 cm
Width (Metric) 10 cm