Learn More
Thermo Scientific™ Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Brand: Thermo Scientific™ B-LP
1251.02 NOK valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
- Size: 36 x 46cm
- Includes bubble level
Specifications
Owl System | |
Owl | |
Leveling Platform |