Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Product Code. 12454637 Shop All Thermo Scientific Products
Change view
Click to view available options
Gel Size:
14 x 14 cm
24 x 24 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Size Unit Size
12454637 14 x 14 cm Each
11560435 24 x 24 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12454637 Supplier Thermo Scientific™ Supplier No. GDF10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
TRUSTED_SUSTAINABILITY
Specifications
Gel Size 14 x 14 cm
Quantity Each
Length (Metric) 14 cm
Width (Metric) 14 cm
Product Type Gel Drying Kit