Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs
Product Code. 12414617
Change view
Click to view available options
Teeth Count:
17
Thickness (Metric) Teeth:
1 mm
1.5 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Teeth Count Thickness (Metric) Teeth unitSize
12414617 17 1 mm Each
12424617 17 1.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12414617 Supplier Thermo Scientific™ Supplier No. D417C
  • ONLINE EXCLUSIVE PRICE
    757.00 NOK
    733.51 NOK / Pack of 1
    Save 23.49 NOK
    3% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Eliminate comb assembly with heavy-duty, one-piece microwell Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs.
Eliminate comb assembly with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs. These heavy-duty, one-piece microwell combs are for use with the Owl D4 Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
TRUSTED_SUSTAINABILITY
Specifications
Description Micro well comb
For Use With (Equipment) Owl™ D4 Electrophoresis System
Product Line Owl
Type Comb
Teeth Count 17
Thickness (Metric) Teeth 1 mm