Learn More
Thermo Scientific™ Owl™ Casting Dams
Shop All Thermo Scientific Products
Click to view available options
Depth (Metric):
13 cm
20 cm
23 cm
Unit Size:
Each
Description
Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.
- Seal agarose on contact, eliminating the need for tape
- Does not attach to gel tray, enabling gels of any length to be poured
- Easily handled and maneuvered using knob or finger depressions
- Dam blocks off a portion of UVT gel tray
Specifications
Specifications
| Description | Casting Dam, 20cm (L) |
| For Use With (Equipment) | Owl System |
| Product Line | Owl |
| Type | Gel Casting Dam |
| Depth (Metric) | 20 cm |