Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts
Shop All Thermo Scientific Products
Click to view available options
Type:
Buffer Chamber and Lid
Casting Slides
Gel Casting Tray
Unit Size:
Each
Description
Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts. Parts are for use with the Owl C2-S Micro Electrophoresis System, a system designed for quick restriction fragment analysis.
- Glass slides, 5 x 7.5 cm, are used as the gel tray
- Thin gels can be poured and formed by surface tension directly on the glass slide
Specifications
Specifications
| For Use With (Equipment) | Owl™ C2-S Micro Electrophoresis System |
| Product Line | Owl |
| Type | Gel Casting Tray |