Description
- The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.
Specifications
Specifications
| For Use With (Equipment) | ADJ1 Giraffe™ Adjustable Gel Electrophoresis System |
| Type | Gasket |