Learn More
Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Standard Comb
Shop All Thermo Scientific Products
Click to view available options
Type:
Rapid Load Comb
Teeth Count:
25 (2X)
9 (1X)
Thickness (Metric) Teeth:
1.0 mm
1.5 mm
Unit Size:
Each
Description
Eliminate comb assembly with this assortment of heavy-duty, one-piece Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Combs, for use with Owl A1 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Specifications
| For Use With (Equipment) | Owl A1 Large Gel Electrophoresis System |
| Product Line | Owl |
| Type | Rapid Load Comb |
| Teeth Count | 9 (1X) |
| Thickness (Metric) Teeth | 1.5 mm |