Learn More
KleenGuard™ A40 Light Duty Overshoe with Sole
Product Code. 12685576
Change view
Click to view available options
Size:
Medium/Large
X-Large/2X-Large
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12685576 Medium/Large Pack of 200
12695576 X-Large/2X-Large Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12685576 Supplier KleenGuard™ Supplier No. 98710
  • ONLINE EXCLUSIVE PRICE
    8310.80 NOK
    4172.43 NOK / Pack of 200
    Save 4138.37 NOK
    49% Off
Estimated Shipment: 18-12-2026
Ideal for keeping footwear clean and covered. KleenGuard™ A40 Light Duty Overshoe with Sole reduces contamination in food processing and light industrial areas.
  • Elasticated overshoe with sole
  • Single-use light duty
  • Ideal for keeping footwear clean and covered
  • Reduce contamination in food processing and light industrial areas
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Application) In industry and laboratories.
Material Spundbond Polypropylene
Size Medium/Large
Toe Material Multi-layer Non-woven Barrier Laminate
Waterproof Water Resistant
Product Type Overshoe