Learn More
Eagle™ Overpacks with Screw-On Lid
Product Code. 17782308
Change view
Click to view available options
Capacity (Metric):
247 L
361 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17782308 361 L Each
17772308 247 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17782308 Supplier Eagle™ Supplier No. EDR16900YL
  • ONLINE EXCLUSIVE PRICE
    5401.15 NOK
    4394.19 NOK / Pack of 1
    Save 1006.96 NOK
    18% Off
Estimated Shipment: 21-12-2026
Eagle Overpacks are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.
  • Eagle Overpack Poly Drums are constructed of yellow blow-moulded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, weather and chemical resistant, and has a yellow screw-on lid
  • Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
  • Overpacks - for use with larger non-damaged containers, can and drums of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
Specifications
Capacity (Metric) 361 L
Color Yellow
For Use With (Application) Safe Storage, Handling and Transportation of Hazardous Materials
Material Polyethylene