Learn More
BINDER™ Overheating Alarm Function for Drying and Heating Chambers
The Overheating Alarm Function for Drying and Heating Chambers features a switchable acoustic alarm, with adjustable limit value on the independent temperature safety device.
Brand:  BINDER™ 8012-1300
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 16444169
  • 8705.00 NOK / Each
Estimated Shipment: 15-07-2024
Description
  • Audible alarm function for overheating
  • Features switchable acoustic alarm
  • Features adjustable limit value
  • Designed for use with BINDER™ Drying and Heating Chambers
Specifications
Specifications
Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device
BINDER Chambers FD 23 Drying and Heating Chambers with Forced Convection, 230 V Model
Adjustable Overheating Alarm
230 V