Learn More
Caframo™ Overhead Stirrer Clamp
Product Code. 16853953
Change view
Click to view available options
Length (Metric):
81 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
16853953 81 mm Each
1 options
Product Code. 16853953
Brand: Caframo™ A122
Only null left
Add to basket
Add to basket
Overhead Stirrer Clamp designed to accommodate stirrer support rods up to 16 mm (5/8 in.) diameter. Attaches overhead stirrer to Caframo and Fisherbrand stands (14-500-354 and 14-500-355).
  • This clamp is an alternative to the A120 and A120FSR (14-500-6, 14-500-357)
  • Compatible with larger diameter support rods, up to 16 mm (5/8 in.)
Specifications
For Use With (Equipment) Caframo™ and Fisherbrand™ Stands (14-500-354 and 14-500-355)
Includes Key Only
Length (Metric) 81 mm
Type Clamp
Width (Metric) 40 mm
Material Cast Zinc, Aluminum