Learn More
Aqualytic™ Overflow Measuring Flask
Product Code. 10079711
Change view
Click to view available options
Volume (Metric):
428 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10079711 428 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10079711 Supplier Aqualytic™ Supplier No. 418660
  • ONLINE EXCLUSIVE PRICE
    592.00 NOK
    547.22 NOK / Pack of 1
    Save 44.78 NOK
    7% Off
Estimated Shipment: 29-01-2027
Aqualytic™ Overflow Measuring Flask is a suitable accessory for BD 600 and BD 606 respirometric method oxygen consumption measuring systems.
  • Suitable for BD 600 or BD 606 respirometric biochemical oxygen demand measuring system
  • Large volume flask
Specifications
Description Overflow Flask
Volume (Metric) 428 mL