Learn More
Labcon Overfit Caps for Culture Tubes
Product Code. 17284406
Click to view available options
Color:
Assorted (Red, Yellow, Blue, Green, Orange)
Blue
Green
Natural
Orange
Purple
Red
Yellow
Closure Size:
10 mm
12 mm
13 mm
16 mm
Unit Size:
Pack of 1000
Pack of 10000
Product Code. 17284406
Brand: Labcon 33038030009
Only null left
Add to basket
Add to basket
Secure, liquid-tight seal for plastic and glass culture tubes. Available in sizes that fit 10 mm, 12 mm, 13 mm, and 16 mm round bottom culture tubes.
  • Designed to easily recap blood drawing tubes, glass test tubes, and plastic culture tubes, Overfit Caps are ideal for protocols that require centrifugation, agitation,
    and refrigeration.
  • Offered in a variety of bright colors, Labcon's Overfit Caps are the ideal alternative to costly parafilm or plug caps that are prone to evaporation.
  • Cap includes integral thumb lever for easy opening and vertical ridges for secure grip
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Closure Size 10 mm
Pre-sterilized No
Product Type Overfit Cap
Autoclavable Not Autoclavable
Material Polyethylene USP Class VI
Color Orange
For Use With (Application) Protocols that require centrifugation, agitation, and refrigeration
Packaging 1000/Bag, 10 Bags/Cs.
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Sterility Non-sterile
For Use With (Equipment) Culture Tubes