Learn More
- Home
- Products
- Tubes
- Tube Caps and Closures
- Labcon Overfit Caps for Culture Tubes
Labcon Overfit Caps for Culture Tubes
Product Code. 17274406
Change view
Click to view available options
Color:
Assorted (Red, Yellow, Blue, Green, Orange)
Blue
Green
Natural
Orange
Purple
Red
Yellow
Closure Size:
10 mm
12 mm
13 mm
16 mm
Unit Size:
Pack of 1000
Pack of 10000
| Product Code. | Color | Closure Size | unitSize |
|---|---|---|---|
| 17274406 | Yellow | 10 mm | Pack of 1000 |
| 17294406 | Red | 10 mm | Pack of 1000 |
| 17284406 | Orange | 10 mm | Pack of 1000 |
| 17224416 | Green | 10 mm | Pack of 1000 |
| 17214416 | Blue | 10 mm | Pack of 1000 |
| 17204416 | Purple | 10 mm | Pack of 1000 |
| 17264406 | Natural | 10 mm | Pack of 1000 |
| 17234416 | Assorted (Red, Yellow, Blue, Green, Orange) | 10 mm | Pack of 1000 |
| 17274416 | Red | 12 mm | Pack of 1000 |
| 17264416 | Orange | 12 mm | Pack of 1000 |
| 17254416 | Yellow | 12 mm | Pack of 1000 |
| 17204426 | Green | 12 mm | Pack of 1000 |
| 17294416 | Blue | 12 mm | Pack of 1000 |
| 17284416 | Purple | 12 mm | Pack of 1000 |
| 17244416 | Natural | 12 mm | Pack of 1000 |
| 17214426 | Assorted (Red, Yellow, Blue, Green, Orange) | 12 mm | Pack of 1000 |
| 17254426 | Red | 13 mm | Pack of 1000 |
| 17244426 | Orange | 13 mm | Pack of 1000 |
| 17234426 | Yellow | 13 mm | Pack of 1000 |
| 17284426 | Green | 13 mm | Pack of 1000 |
| 17567606 | Blue | 13 mm | Pack of 10000 |
| 17264426 | Purple | 13 mm | Pack of 1000 |
| 17224426 | Natural | 13 mm | Pack of 1000 |
| 17294426 | Assorted (Red, Yellow, Blue, Green, Orange) | 13 mm | Pack of 1000 |
| 17234436 | Red | 16 mm | Pack of 1000 |
| 17224436 | Orange | 16 mm | Pack of 1000 |
| 17214436 | Yellow | 16 mm | Pack of 1000 |
| 17264436 | Green | 16 mm | Pack of 1000 |
| 17254436 | Blue | 16 mm | Pack of 1000 |
| 17244436 | Purple | 16 mm | Pack of 1000 |
| 17204436 | Natural | 16 mm | Pack of 1000 |
| 17274436 | Assorted (Red, Yellow, Blue, Green, Orange) | 16 mm | Pack of 1000 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Designed to easily recap blood drawing tubes, glass test tubes, and plastic culture tubes, Overfit Caps are ideal for protocols that require centrifugation, agitation,
and refrigeration. - Offered in a variety of bright colors, Labcon's Overfit Caps are the ideal alternative to costly parafilm or plug caps that are prone to evaporation.
- Cap includes integral thumb lever for easy opening and vertical ridges for secure grip
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polyethylene USP Class VI resin
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
| Closure Size | 10 mm |
| Pre-sterilized | No |
| Product Type | Overfit Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene USP Class VI |
| Color | Yellow |
| For Use With (Application) | Protocols that require centrifugation, agitation, and refrigeration |
| Packaging | 1000/Bag, 10 Bags/Cs. |
| Clean Claims | ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) |
| Sterility | Non-sterile |
| For Use With (Equipment) | Culture Tubes |
| Disposable | Single-use |
| Show More Show Less |