Learn More
OVCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14943-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16883864
  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
Description
Chromosome 17 is a hotspot for chromosomal aberrations in breast and ovarian cancer. Candidate oncogenes and tumor suppressors located on this chromosome include OVCA1 and OVCA2, p53, BRCA1, HER2/neu and others. OVCA1 and 2 map to a highly conserved region on human chromosome 17p13.3 that is deleted in 80% of ovarian cancers and shows frequent loss of heterozygosity in breast cancers. OVCA1 is induced by BRCA1 and is closely linked to p53, a well-known tumor suppressor gene. The close linkage of OVCA1 and p53 suggests that coordinated loss of the two genes may lead to ovarian, breast and other tumor types.
Specifications
Specifications
OVCA2
Polyclonal
Unconjugated
OVCA2
OVCA2
Rabbit
Antigen Affinity Chromatography
RUO
124641, 246257, 497954
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WZ82, Q9D7E3
Ovca2
OVCA2 Fusion Protein Ag6744
20 μL
Primary
Human, Rat, Mouse