Learn More
OVCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14943-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Chromosome 17 is a hotspot for chromosomal aberrations in breast and ovarian cancer. Candidate oncogenes and tumor suppressors located on this chromosome include OVCA1 and OVCA2, p53, BRCA1, HER2/neu and others. OVCA1 and 2 map to a highly conserved region on human chromosome 17p13.3 that is deleted in 80% of ovarian cancers and shows frequent loss of heterozygosity in breast cancers. OVCA1 is induced by BRCA1 and is closely linked to p53, a well-known tumor suppressor gene. The close linkage of OVCA1 and p53 suggests that coordinated loss of the two genes may lead to ovarian, breast and other tumor types.Specifications
OVCA2 | |
Polyclonal | |
Unconjugated | |
OVCA2 | |
OVCA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124641, 246257, 497954 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WZ82, Q9D7E3 | |
Ovca2 | |
OVCA2 Fusion Protein Ag6744 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |