Learn More
VistaLab™ Technologies Replacement Duracell™ Ultra CR2 Battery for Ovation™ QS Pipette
Product Code. 11955962
Change view
Click to view available options
Battery Type:
CR2032
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Type Unit Size
11955962 CR2032 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11955962 Supplier VistaLab™ Technologies Supplier No. 90574001
  • ONLINE EXCLUSIVE PRICE
    294.00 NOK
    227.11 NOK / Pack of 1
    Save 66.89 NOK
    22% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
3 volt lithium battery with an expected life of one year or more of typical use.
Specifications
Product Type CR2 Battery
Battery Type CR2032
For Use With (Equipment) Ovation Adjustable Volume Pipettes
Quantity 1 Ea.
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.
Voltage 3 V