Learn More
R&D Systems™ Ovary Tissue Slides (Dermoid Cyst)- Paraffin
Product Code. 18109583 Shop All Bio Techne Products
Change view
Click to view available options
Species:
Human
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species Unit Size
18109583 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18109583 Supplier R&D Systems™ Supplier No. NBP230291
 Restricted Item
Estimated Shipment: 11-12-2026
A Novus product, part of R&D Systems.
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
TRUSTED_SUSTAINABILITY
Specifications
Cross Reactivity Human
Type Ovary Tissue Slides (Dermoid Cyst)
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Dermoid Cyst
Product Line Novus