Learn More
Cowie Technology™ Oval PTFE Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
70 mm
100 mm
150 mm
Diameter (Metric):
25 mm
30 mm
40 mm
Unit Size:
Each
Description
- Stable, general-purpose magnetic stir bar
- Provides excellent turbulence for round-bottom flasks and containers
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Round bottom flasks |
| Diameter (Metric) | 25 mm |
| Length (Metric) | 70 mm |
| Quantity | 1/Ea. |
| Shape | Oval |