missing translation for 'onlineSavingsMsg'
Learn More

OTS514, MedChemExpress

Product Code. p-200096673
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16487538 10 mg 10mg
16470018 5 mg 5mg
16467486 100 mg 100mg
16463606 25 mg 25mg
16430306 2 mg 2mg
16403567 50 mg 50mg
6 options
This item is not returnable. View return policy

Product Code. 16487538

Brand: Medchem Express HY18621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OTS514 is a highly potent TOPK inhibitor with an IC50 of 2.6 nM. OTS514 strongly suppresses the growth of TOPK-positive cancer cells. OTS514 induces cell cycle arrest and apoptosis.

OTS514 is a highly potent TOPK inhibitor with an IC50 of 2.6 nM. OTS514 strongly suppresses the growth of TOPK-positive cancer cells. OTS514 induces cell cycle arrest and apoptosis.

Specifications

Chemical Name or Material OTS514
CAS 1338540-63-8
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C21H20N2O2S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (274.38 mM; Need ultrasonic)
SMILES O=C1NC2=C(C(C3=CC=C([C@@H](C)CN)C=C3)=C(O)C=C2C)C4=C1SC=C4
Molecular Weight (g/mol) 364.46
Formula Weight 364.46
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.