missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OTOR Polyclonal Antibody

Product Code. 15732981 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15732981 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15732981 Supplier Invitrogen™ Supplier No. PA555053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RLASKKLCAD DECVYTISLA SAQEDYNAPD CRFINVKKGQ QIYVYSKLVK ENGAGEFWAG SVYGDGQDEM GVVGYFPRNL VKEQRVYQEA TKEVPTTDID FFCE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.

OTOR, also called Otoraplin and MIAL, is a secreted cytokine of the MIA/OTOR family. Members of this family, which include MIA, MIA2, and TANGO, share a Src homology-3 (SH3)-like domain. OTOR is predominantly expressed in the cochlea of the inner ear, and, to a lesser extent, in fetal brain tissue and some cartilage tissues. OTOR appears to be involved in early chondrogenesis of the otic capsule, which is required for normal inner ear development and auditory function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTOR
Gene Accession No. Q9NRC9
Gene Alias CDRAP; FDP; Fibrocyte-derived protein; melanoma inhibitory activity-like protein; MIA; MIAL; MIAL1; Otor; Otoraplin; UNQ3054/PRO9873
Gene Symbols OTOR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTOR. Recombinant protein control fragment (Product #RP-90188).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.