Learn More
Descrizione
- Osmolality Reference Standard, Osmolality Reference Material.
- The Osmolality results are reported in milliOsmoles per kilogram water (mOsm/kg H2O shortened to mOsm/kg for routine use).
- This product is manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines where appropriate.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref 264T).
- Prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
- Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The certified values of each standard are verified using a high-performance osmometer calibrated with high quality reference materials.
Specifica
Specifica
| Packaging | Amber Glass Ampule |
| Physical Form | Liquid |
| Concentration | 3000 mOsm/Kg |
| Technique | Calibration of Osmometer |
| Quantity | 12 x 5 mL |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?