missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OSBPL3 Polyclonal antibody specifically detects OSBPL3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | OSBPL3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp667P1518, KIAA0704ORP3ORP-3OSBP3, MGC21526, OSBP-related protein 3, oxysterol binding protein-like 3, oxysterol-binding protein 3, oxysterol-binding protein-related protein 3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 370-520 of human OSBPL3 (NP_056365.1).,, Sequence:, SSSPSAQVIGLKNALSSALAQNTDLKERLRRIHAESLLLDSPAVAKSGDNLAEENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPC |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?