Learn More
Thermo Scientific™ Orion™ All-in-One™ pH Buffer Kits
Product Code. p-10062000
Change view
Click to view available options
Description:
All-in-One pH Buffer Kit
ROSS All-in-One pH Buffer Kit
Standard All-in-One pH Buffer Kit
Unit Size:
475mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10669508 ROSS All-in-One pH Buffer Kit 475mL
12973247 All-in-One pH Buffer Kit 60mL
10432062 Standard All-in-One pH Buffer Kit 475mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10669508 Supplier Thermo Scientific™ Supplier No. 810199
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
Kit includes one each:
  • 1 pint (475mL) bottle of pH 4.01 buffer (910104)
  • 1 pint (475mL) bottle of pH 7.00 buffer (910107)
  • 1 pint (475mL) bottle of pH 10.01 buffer (910110)
  • 1 pint (475mL) bottle of ROSS™ pH electrode storage solution (810001)
  • 1 oz. (60mL) bottle of general pH electrode cleaning solution (900024)
  • 12mm diameter pH electrode storage bottle
Click here for the most complete, up-to-date MSDS and certificates of analysis files for Orion solutions
TRUSTED_SUSTAINABILITY
Specifications
Description ROSS All-in-One pH Buffer Kit
For Use With (Application) Accurate calibrations and NIST traceability, laboratory and bench analysis
pH 4.01, 7.00, 10.01
Quantity 4 x 475 mL, 1 x 60 mL
Solution Type pH Buffer, Storage Solution and Cleaning Solution
Packaging Kit
Traceability to NIST Yes
Includes 475 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 475 mL bottle of ROSS pH electrode storage solution (810001); 60 mL bottle of pH electrode cleaning solution; and 12 mm diameter electrode storage bottle
Product Identifier
  • Thermo Scientific Orion ROSS All-in-One pH Buffer Kit
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P202-Do not handle until all safety precautions have been read and understood.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Lauryl Sulfate